Imprimer

Anti-LGALS3BP Mouse Monoclonal Antibody [clone: OTI5E3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503457EA 603 EUR
ORIGTA503457
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: OTI5E3]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LGALS3BP (lectin, galactoside-binding, soluble, 3 binding protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR