Imprimer
Anti-UNG Mouse Monoclonal Antibody [clone: OTI1A6]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA503502EA 603 EUR
ORIGTA503502
Anti-UNG Mouse Monoclonal Antibody [clone: OTI1A6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: UNG (uracil-DNA glycosylase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human