Imprimer

Anti-CTBP1-AS2 Mouse Monoclonal Antibody [clone: OTI4F2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503514EA 608 EUR
ORIGTA503514
Anti-CTBP1-AS2 Mouse Monoclonal Antibody [clone: OTI4F2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CTBP1-AS2 (CTBP1 antisense RNA 2 (head to head))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR