Imprimer
Anti-ERBB2 Mouse Monoclonal Antibody [clone: OTI1C5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA503523EA 603 EUR
ORIGTA503523
Anti-ERBB2 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human