Imprimer

Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1H1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503570EA 603 EUR
ORIGTA503570
Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1H1]
Anticorps
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: MTFMT (mitochondrial methionyl-tRNA formyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR