Imprimer

Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503611EA 603 EUR
ORIGTA503611
Anti-ACSS2 Mouse Monoclonal Antibody [clone: OTI2E1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ACSS2 (acyl-CoA synthetase short-chain family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR