Imprimer

Anti-TMED1 Mouse Monoclonal Antibody [clone: OTI7D2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503620EA 608 EUR
ORIGTA503620
Anti-TMED1 Mouse Monoclonal Antibody [clone: OTI7D2]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: TMED1 (transmembrane emp24 protein transport domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR