Imprimer
Anti-TP73-AS1 Mouse Monoclonal Antibody [clone: OTI2B9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA503633EA 581 EUR
ORIGTA503633
Anti-TP73-AS1 Mouse Monoclonal Antibody [clone: OTI2B9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: TP73-AS1 (TP73 antisense RNA 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human