Imprimer

Anti-RPN1 Mouse Monoclonal Antibody [clone: OTI2G1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503654EA 603 EUR
ORIGTA503654
Anti-RPN1 Mouse Monoclonal Antibody [clone: OTI2G1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RPN1 (ribophorin I)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR