Imprimer

Anti-TP53I3 Mouse Monoclonal Antibody [clone: OTI3B11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503656EA 586 EUR
ORIGTA503656
Anti-TP53I3 Mouse Monoclonal Antibody [clone: OTI3B11]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TP53I3 (tumor protein p53 inducible protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR