Imprimer

Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI3E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503663EA 603 EUR
ORIGTA503663
Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI3E1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NIF3L1 (NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR