Imprimer

Anti-TBC1D21 Mouse Monoclonal Antibody [clone: OTI3C2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503719EA 581 EUR
ORIGTA503719
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: OTI3C2]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: TBC1D21 (TBC1 domain family, member 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR