Imprimer

Anti-NUDT18 Mouse Monoclonal Antibody [clone: OTI1A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503830EA 581 EUR
ORIGTA503830
Anti-NUDT18 Mouse Monoclonal Antibody [clone: OTI1A5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NUDT18 (nudix (nucleoside diphosphate linked moiety X)-type motif 18)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR