Imprimer

Anti-CYP2A6 Mouse Monoclonal Antibody [clone: OTI1D2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503857EA 608 EUR
ORIGTA503857
Anti-CYP2A6 Mouse Monoclonal Antibody [clone: OTI1D2]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP2A6 (cytochrome P450, family 2, subfamily A, polypeptide 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR