Imprimer

Anti-TFG Mouse Monoclonal Antibody [clone: OTI2C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA503858
ORIGTA503858EA 621 EUR
ORIGTA503858
Anti-TFG Mouse Monoclonal Antibody [clone: OTI2C3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TFG (TRK-fused gene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR