Imprimer

Anti-CDH2 Mouse Monoclonal Antibody [clone: OTI2G7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503933EA 608 EUR
ORIGTA503933
Anti-CDH2 Mouse Monoclonal Antibody [clone: OTI2G7]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CDH2 (cadherin 2, type 1, N-cadherin (neuronal))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR