Imprimer

Anti-C1S Mouse Monoclonal Antibody [clone: OTI1G1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA503953EA 581 EUR
ORIGTA503953
Anti-C1S Mouse Monoclonal Antibody [clone: OTI1G1]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR