Imprimer

Anti-SERPINB6 Mouse Monoclonal Antibody [clone: OTI2F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504055EA 603 EUR
ORIGTA504055
Anti-SERPINB6 Mouse Monoclonal Antibody [clone: OTI2F8]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SERPINB6 (serpin peptidase inhibitor, clade B (ovalbumin), member 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR