Imprimer
Anti-NMT2 Mouse Monoclonal Antibody [clone: OTI2E1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA504072EA 581 EUR
ORIGTA504072
Anti-NMT2 Mouse Monoclonal Antibody [clone: OTI2E1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: NMT2 (N-myristoyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human