Imprimer

Anti-MKRN1 Mouse Monoclonal Antibody [clone: OTI2C8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504092EA 586 EUR
ORIGTA504092
Anti-MKRN1 Mouse Monoclonal Antibody [clone: OTI2C8]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MKRN1 (makorin ring finger protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR