Imprimer
Anti-IGJ Mouse Monoclonal Antibody [clone: OTI3E12]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA504172EA 603 EUR
ORIGTA504172
Anti-IGJ Mouse Monoclonal Antibody [clone: OTI3E12]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: IGJ (immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human