Imprimer

Anti-AGPAT5 Mouse Monoclonal Antibody [clone: OTI1D12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504192EA 608 EUR
ORIGTA504192
Anti-AGPAT5 Mouse Monoclonal Antibody [clone: OTI1D12]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: AGPAT5 (1-acylglycerol-3-phosphate O-acyltransferase 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR