Imprimer

Anti-CAPN2 Mouse Monoclonal Antibody [clone: OTI1F4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504345EA 608 EUR
ORIGTA504345
Anti-CAPN2 Mouse Monoclonal Antibody [clone: OTI1F4]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CAPN2 (calpain 2, (m/II) large subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR