Imprimer

Anti-DPP8 Mouse Monoclonal Antibody [clone: OTI1D2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504366EA 621 EUR
ORIGTA504366
Anti-DPP8 Mouse Monoclonal Antibody [clone: OTI1D2]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150

Type: Primary
Antigen: DPP8 (dipeptidyl-peptidase 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR