Imprimer

Anti-PSMB7 Mouse Monoclonal Antibody [clone: OTI4G9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504383EA 586 EUR
ORIGTA504383
Anti-PSMB7 Mouse Monoclonal Antibody [clone: OTI4G9]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMB7 (proteasome (prosome, macropain) subunit, beta type, 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR