Imprimer

Anti-MPP5 Mouse Monoclonal Antibody [clone: OTI5D10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504458EA 581 EUR
ORIGTA504458
Anti-MPP5 Mouse Monoclonal Antibody [clone: OTI5D10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: MPP5 (membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR