Imprimer

Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504493EA 603 EUR
ORIGTA504493
Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C11]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR