Imprimer

Anti-ATIC Mouse Monoclonal Antibody [clone: OTI4A10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504570EA 603 EUR
ORIGTA504570
Anti-ATIC Mouse Monoclonal Antibody [clone: OTI4A10]
Anticorps
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: ATIC (5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR