Imprimer

Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI4H3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504578EA 603 EUR
ORIGTA504578
Anti-DNAAF1 Mouse Monoclonal Antibody [clone: OTI4H3]
Anticorps
  • Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: DNAAF1 (dynein, axonemal, assembly factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR