Imprimer

Anti-SERPINE2 Mouse Monoclonal Antibody [clone: OTI3B12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504602EA 603 EUR
ORIGTA504602
Anti-SERPINE2 Mouse Monoclonal Antibody [clone: OTI3B12]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINE2 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR