Imprimer

Anti-CRYZL1 Mouse Monoclonal Antibody [clone: OTI1C9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504607EA 603 EUR
ORIGTA504607
Anti-CRYZL1 Mouse Monoclonal Antibody [clone: OTI1C9]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CRYZL1 (crystallin, zeta (quinone reductase)-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR