Imprimer

Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1D1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504610EA 603 EUR
ORIGTA504610
Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1D1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FXYD3 (FXYD domain containing ion transport regulator 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR