Imprimer

Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1D4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504626EA 603 EUR
ORIGTA504626
Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1D4]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: MIEN1 (migration and invasion enhancer 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR