Imprimer

Anti-SCFD1 Mouse Monoclonal Antibody [clone: OTI1E2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504667EA 603 EUR
ORIGTA504667
Anti-SCFD1 Mouse Monoclonal Antibody [clone: OTI1E2]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SCFD1 (sec1 family domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR