Imprimer
Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI2E7]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA504676EA 581 EUR
ORIGTA504676
Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI2E7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: CPA2 (carboxypeptidase A2 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human