Imprimer

Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI3D8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504696EA 581 EUR
ORIGTA504696
Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI3D8]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:150

Type: Primary
Antigen: CPA2 (carboxypeptidase A2 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR