Imprimer

Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2H10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504766EA 603 EUR
ORIGTA504766
Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2H10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MLF1 (myeloid leukemia factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR