Imprimer

Anti-VBP1 Mouse Monoclonal Antibody [clone: OTI1A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504894EA 603 EUR
ORIGTA504894
Anti-VBP1 Mouse Monoclonal Antibody [clone: OTI1A5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: VBP1 (von Hippel-Lindau binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR