Imprimer

Anti-NRIP3 Mouse Monoclonal Antibody [clone: OTI1G6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA504921EA 603 EUR
ORIGTA504921
Anti-NRIP3 Mouse Monoclonal Antibody [clone: OTI1G6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NRIP3 (nuclear receptor interacting protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR