Imprimer
Anti-ESRRG Mouse Monoclonal Antibody [clone: OTI1D10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGTA504963EA 603 EUR
ORIGTA504963
Anti-ESRRG Mouse Monoclonal Antibody [clone: OTI1D10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: ESRRG (estrogen-related receptor gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human