Imprimer

Anti-MMACHC Mouse Monoclonal Antibody [clone: OTI4E12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA504998
ORIGTA504998EA 593 EUR
ORIGTA504998
Anti-MMACHC Mouse Monoclonal Antibody [clone: OTI4E12]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MMACHC (methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR