Imprimer

Anti-MINOS1 Mouse Monoclonal Antibody [clone: OTI4H7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505025EA 609 EUR
ORIGTA505025
Anti-MINOS1 Mouse Monoclonal Antibody [clone: OTI4H7]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: MINOS1 (mitochondrial inner membrane organizing system 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR