Imprimer

Anti-CBLC Mouse Monoclonal Antibody [clone: OTI3B4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505071EA 608 EUR
ORIGTA505071
Anti-CBLC Mouse Monoclonal Antibody [clone: OTI3B4]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CBLC (Cbl proto-oncogene C, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR