Imprimer

Anti-LEPRE1 Mouse Monoclonal Antibody [clone: OTI1H3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505088EA 608 EUR
ORIGTA505088
Anti-LEPRE1 Mouse Monoclonal Antibody [clone: OTI1H3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LEPRE1 (leucine proline-enriched proteoglycan (leprecan) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR