Imprimer

Anti-TRIM22 Mouse Monoclonal Antibody [clone: OTI1A10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505295EA 603 EUR
ORIGTA505295
Anti-TRIM22 Mouse Monoclonal Antibody [clone: OTI1A10]
Anticorps
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TRIM22 (tripartite motif containing 22)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR