Imprimer

Anti-RAB30 Mouse Monoclonal Antibody [clone: OTI3B7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505316EA 616 EUR
ORIGTA505316
Anti-RAB30 Mouse Monoclonal Antibody [clone: OTI3B7]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: RAB30 (RAB30, member RAS oncogene family)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR