Imprimer

Anti-ETS2 Mouse Monoclonal Antibody [clone: OTI1H4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505372EA 586 EUR
ORIGTA505372
Anti-ETS2 Mouse Monoclonal Antibody [clone: OTI1H4]
Anticorps
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: ETS2 (v-ets avian erythroblastosis virus E26 oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR