Imprimer

Anti-CATIP Mouse Monoclonal Antibody [clone: OTI2C6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505505EA 581 EUR
ORIGTA505505
Anti-CATIP Mouse Monoclonal Antibody [clone: OTI2C6]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CATIP (ciliogenesis associated TTC17 interacting protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR