Imprimer

Anti-CRYAA Mouse Monoclonal Antibody [clone: OTI2H2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505529EA 603 EUR
ORIGTA505529
Anti-CRYAA Mouse Monoclonal Antibody [clone: OTI2H2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CRYAA (crystallin, alpha A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR