Imprimer

Anti-GBAS Mouse Monoclonal Antibody [clone: OTI1B8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505590EA 586 EUR
ORIGTA505590
Anti-GBAS Mouse Monoclonal Antibody [clone: OTI1B8]
Anticorps
  • Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: GBAS (glioblastoma amplified sequence)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR